missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, TRC

Product Code. 30410402
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410402 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410402 Supplier Toronto Research Chemicals Supplier No. TRCP3360485G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9 H12 O
Molecular Weight (g/mol) 136.19
Synonym Benzenemethanol, α,α-dimethyl-, 2-Phenyl-2-propanol, Benzyl alcohol, α,α-dimethyl- (8CI), α,α-Dimethylbenzenemethanol, 1-Hydroxycumene, 1-Methyl-1-phenylethanol, 2-Hydroxy-2-phenylpropane, 2-Phenyl-2-propanol, 2-Phenylisopropanol, 2-Propanol, 2-phenyl-, Dimethylphenylcarbinol, Dimethylphenylmethanol, NSC 1261, NSC 212537, Phenyldimethylcarbinol, α,α-Dimethylbenzyl alcohol, α-Cumyl alcohol
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)c1ccccc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H12O/c1-9(2,10)8-6-4-3-5-7-8/h3-7,10H,1-2H3
Formula Weight 136.0888
Chemical Name or Material 2-Phenyl-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.