missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, MedChemExpress

Product Code. 16462329
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16462329 Supplier Medchem Express Supplier No. HYB1290
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress 2-Phenylethanol (Phenethyl alcohol), extracted from rose, carnation, hyacinth, Aleppo pine, orange blossom and other organisms, is a colourless liquid. It has a pleasant floral odor and also an autoantibiotic produced by the fungus Candida albicans. It is used as an additive in cigarettes and also used as a preservative in soaps due to its stability in basic conditions.

2-Phenylethanol (Phenethyl alcohol), extracted from rose, carnation, hyacinth, Aleppo pine, orange blossom and other organisms, is a colourless liquid. It has a pleasant floral odor and also an autoantibiotic produced by the fungus Candida albicans. It is used as an additive in cigarettes and also used as a preservative in soaps due to its stability in basic conditions.

Specifications

Chemical Name or Material 2-Phenylethanol
Physical Form Liquid
CAS 60-12-8
Color Colorless
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H10O
For Use With (Application) COVID-19-anti-virus
Synonym Phenylethyl alcohol Phenethyl alcohol Benzyl carbinol
Solubility Information H2O : 20 mg/mL (163.72 mM; Need ultrasonic)
SMILES OCCC1=CC=CC=C1
Molecular Weight (g/mol) 122.16
Formula Weight 122.16
Percent Purity 99.64%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.