missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylnaphthalene, 98%, Thermo Scientific™

Product Code. 15239305 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15239305 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15239305 Supplier Thermo Scientific Acros Supplier No. 450070010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 612-94-2
Melting Point 105°C
Boiling Point 358°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C16H12
Quantity 1 g
Synonym naphthalene, 2-phenyl, .beta.-phenylnaphthalene, unii-949vn5dh7a, beta-phenylnaphthalene, 6-phenylnaphthalene, 2-phenyl-naphthalene, acmc-1bas3, naphthalene, 2-phenyl-8ci 9ci, 10 microg/ml in cyclohexane
Solubility Information Solubility in water: insoluble
InChI Key TURIHPLQSRVWHU-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2
IUPAC Name 2-phenylnaphthalene
Molecular Weight (g/mol) 204.27
PubChem CID 11934
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material 2-Phenylnaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.