missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylphenol, Dr. Ehrenstorfer

Product Code. 30311742
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311742 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311742 Supplier Dr. Ehrenstorfer Supplier No. DREC16070000100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenylphenol
CAS 90-43-7
Analyte or Component Names 2-Phenylphenol
Shipping Condition +20°C
Molecular Formula C12 H10 O
Certifications/Compliance ISO 17034
Synonym [1,1'-Biphenyl]-2-ol, 2-Biphenylol (8CI), 2-Hydroxy-1,1'-biphenyl, 2-Hydroxybiphenyl, 2-Hydroxydiphenyl, 2-Phenylphenol, Anthrapole 73, Britewood S, Decco OPP 20, Dowicide 1, Invalon OP, NSC 1548, Nectryl, O-PP, Preventol 3041, Preventol O extra, Remol TRF, Tetrosin OE, Tetrosin OE-N, Tumescal OPE, o-Biphenylol, o-Diphenylol, o-Hydroxybiphenyl, o-Hydroxydiphenyl, o-Phenylphenol, o-Xenol
InChI Formula InChI=1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H
SMILES Oc1ccccc1c2ccccc2
IUPAC Name 2-phenylphenol
Molecular Weight (g/mol) 170.21
Quantity 100 mg
Formula Weight 170.0732 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.