missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Phenylpropionic acid, 98%

Product Code. 11300184
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11300184 5 g 5g
11310184 25 g 25g
11320184 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300184 Supplier Thermo Scientific Alfa Aesar Supplier No. L00709.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 492-37-5 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-37-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002650
InChI Key YPGCWEMNNLXISK-UHFFFAOYSA-N
Synonym 2-phenylpropionic acid, hydratropic acid, dl-2-phenylpropionic acid, alpha-methylphenylacetic acid, 2-phenyl-propionic acid, +/--2-phenylpropionic acid, alpha-methylbenzeneacetic acid, benzeneacetic acid, .alpha.-methyl, hydratropasaeure, alpha-phenylpropionic acid
PubChem CID 10296
ChEBI CHEBI:48526
IUPAC Name 2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point -5°C
Density 1.1
Boiling Point 264°C to 266°C
Flash Point 148°C (298°F)
Refractive Index 1.523
Quantity 5 g
Beilstein 1863558
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material (+/-)-2-Phenylpropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.