missing translation for 'onlineSavingsMsg'
Learn More

2-Picoline, 98%

Product Code. 10437920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10437920 1 L Glass bottle 1 L
10336790 25 mL Glass bottle 25mL
10285430 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437920 Supplier Thermo Scientific Acros Supplier No. 149330010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-06-8 | C6H7N | 93.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-06-8
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00006332
InChI Key BSKHPKMHTQYZBB-UHFFFAOYSA-N
Synonym 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha
PubChem CID 7975
ChEBI CHEBI:50415
IUPAC Name 2-methylpyridine
SMILES CC1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Yellow
Density 0.9400g/mL
Boiling Point 128.0°C to 129.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4990 to 1.5010
Quantity 1 L
Beilstein 20, 234
Merck Index 15, 7512
Specific Gravity 0.94
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 93.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Picoline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Picoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.