missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Product Code. 11926061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11926061 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11926061 Supplier Thermo Scientific Acros Supplier No. 444650010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 159624-15-4
Assay Percent Range 97%
Molecular Formula C17H26BNO4
Quantity 1 g
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-n-boc-amino phenylboronic acid pinacol ester, 2-boc-amino benzeneboronic acid pinacol ester, 2-boc-aminophenylboronic acid pinacol ester, 2-boc-aminophenylboronic acid, pinacol ester, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
InChI Key LVHGGVGVAUJQBB-UHFFFAOYSA-N
Molecular Weight (g/mol) 319.21
PubChem CID 2773231
Formula Weight 319.21
Chemical Name or Material 2-(tert-Butoxycarbonylamino)phenylboronic acid pinacol ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.