missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)aniline, 95%, Thermo Scientific™

Product Code. 10143764
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10143764

missing translation for 'mfr': Thermo Scientific Maybridge CC52214CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-35-5
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD09025872
InChI Key CLGPZXHLRKSEDD-UHFFFAOYSA-N
Synonym 2-tetrahydro-2h-pyran-4-yloxy aniline, 2-tetrahydropyran-4-yloxy aniline, 2-tetrahydro-2h-pyran-4-yl oxy aniline, 2-oxan-4-yloxy aniline, benzenamine,2-tetrahydro-2h-pyran-4-yl oxy, benzenamine, 2-tetrahydro-2h-pyran-4-yl oxy, 2-oxan-4-yl oxy aniline, 2-tetrahydropyran-4-yloxyaniline, 4-2-aminophenoxy tetrahydro-2h-pyran, 2-tetrahydro-2h-pyran-4-yl ;oxy ;aniline
PubChem CID 24229516
IUPAC Name 2-(oxan-4-yloxy)aniline
SMILES C1COCCC1OC2=CC=CC=C2N

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.24
Percent Purity 95%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.