missing translation for 'onlineSavingsMsg'
Learn More

[2-(Tetrahydropyran-4-yloxy)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10662315
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10662315 1 g Amber Glass Bottle 1g
11300697 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662315 Supplier Thermo Scientific Maybridge Supplier No. CC52209DA
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 478189-93-4
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD09064960
InChI Key RMRHXMQGVDIPCL-UHFFFAOYSA-N
Synonym 2-tetrahydropyran-4-yloxy phenyl methanol, 2-oxan-4-yloxy phenyl methanol, 2-oxan-4-yl oxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yloxy phenyl methanol, 2-tetrahydro-2h-pyran-4-yl oxy phenyl methanol, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methan-1-ol
PubChem CID 22932192
IUPAC Name [2-(oxan-4-yloxy)phenyl]methanol
SMILES C1COCCC1OC2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material [2-(Tetrahydropyran-4-yloxy)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.