missing translation for 'onlineSavingsMsg'
Learn More

2-Tetralone, 99%

Product Code. 11304434
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11304434 5 g 5g
11314434 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11304434 Supplier Thermo Scientific Alfa Aesar Supplier No. L01053.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 530-93-8 | C10H10O | 146.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Specifications

Melting Point 18°C
Density 1.106
Boiling Point 240°C
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 5 g
Beilstein 509386
Sensitivity Air sensitive
Formula Weight 146.19
Percent Purity 99%
Chemical Name or Material 2-Tetralone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.