missing translation for 'onlineSavingsMsg'
Learn More

(2-Thien-2-ylphenyl)methanol, 97%, Thermo Scientific™

Product Code. 10122804
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10122804 10 g 10g
10411293 250 mg 250mg
10528043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10122804 Supplier Thermo Scientific Maybridge Supplier No. CC35509EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 773872-97-2
Molecular Formula C11H10OS
Molecular Weight (g/mol) 190.26
MDL Number MFCD06203081
InChI Key IUULMJAEIKVTKZ-UHFFFAOYSA-N
Synonym 2-thien-2-ylphenyl methanol, 2-thiophen-2-yl phenyl methanol, 2-thien-2-yl phenyl methanol, 2-thiophen-2-ylphenyl methanol, 2-2-thienyl phenyl methanol, benzenemethanol,2-2-thienyl, 2-2-thienyl phenyl methan-1-ol
PubChem CID 2795559
SMILES OCC1=CC=CC=C1C1=CC=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
IUPAC Name (2-thiophen-2-ylphenyl)methanol
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (2-Thien-2-ylphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.