missing translation for 'onlineSavingsMsg'
Learn More

2-Thionaphthol, 98%

Product Code. 11450073
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450073 50 g 50g
11440073 10 g 10g
11460073 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450073 Supplier Thermo Scientific Alfa Aesar Supplier No. A12125.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-60-1 | C10H8S | 160.234 g/mol

2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thionaphthol was used in the preparation of cholesterol monolayer and multilayer Langmuir- Blodgett (LB) films. The magnetic resonance shift for a self-assembled monolayer of 2-naphthalenethiol was studied that suggested considerable promise in flexible and transparent photonic devices for biological and chemical sensing.

Solubility
Soluble in alcohol, dipropylene glycol. Soluble in water 34.4 mg/L @ 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-60-1
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.234
MDL Number MFCD00004086
InChI Key RFCQDOVPMUSZMN-UHFFFAOYSA-N
Synonym 2-naphthalenethiol, 2-mercaptonaphthalene, 2-thionaphthol, thionaphthol, 2-naphthyl mercaptan, vulcamel tbn, 2-naphthyl thiol, renacit 1, rpa no. 2, rpa 2
PubChem CID 7058
IUPAC Name naphthalene-2-thiol
SMILES C1=CC=C2C=C(C=CC2=C1)S

Specifications

Melting Point 79°C to 83°C
Density 1.55
Boiling Point 286°C
Quantity 50 g
Beilstein 636389
Sensitivity Air Sensitive
Merck Index 14,6379
Solubility Information Soluble in alcohol,dipropylene glycol. Soluble in water 34.4mg/L @ 25°C.
Formula Weight 160.24
Percent Purity ≥98%
Chemical Name or Material 2-Thionaphthol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.