missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 10712777
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10712777 25 g 25g
10463685 50 g 50g
10504704 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10712777 Supplier Thermo Scientific Maybridge Supplier No. SB01054EB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00035959
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CF3OC6H4NH2
Quantity 25 g
Formula Weight 177.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.