missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™

Product Code. p-2696513
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10540553 1 g 1g
10596785 5 g 5g
10653435 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10540553 Supplier Thermo Scientific Maybridge Supplier No. TL00372DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1979-29-9
Molecular Formula C8H4F3O3
Molecular Weight (g/mol) 205.11
MDL Number MFCD00052325
InChI Key JMYSPFGUBNENSE-UHFFFAOYSA-M
Synonym 2-trifluoromethoxy benzoic acid, o-trifluoromethoxybenzoic acid, alpha,alpha,alpha-trifluoro-o-anisic acid, 2-trifluoromethoxy-benzoic acid, rarechem al bo 0449, pubchem4587, acmc-209f2a, ksc494o7t
PubChem CID 2777223
IUPAC Name 2-(trifluoromethoxy)benzoic acid
SMILES [O-]C(=O)C1=CC=CC=C1OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4CO2H
Quantity 1 g
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.