missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzyl alcohol, 97%, Thermo Scientific™

Product Code. 11489766
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11489766

Brand: Thermo Scientific Alfa Aesar B20274.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-07-6
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.14
MDL Number MFCD00153285
InChI Key ICOVMLDFMWLRJO-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzyl alcohol, 2-trifluoromethoxy phenyl methanol, 2-trifluoromethoxy benzylalcohol, 2-trifluoromethoxybenzyl alcohol, benzenemethanol, 2-trifluoromethoxy, 2-trifluoromethoxy phenyl methan-1-ol, pubchem2745, acmc-209e9m, rarechem al bd 0282
PubChem CID 2777245
IUPAC Name [2-(trifluoromethoxy)phenyl]methanol
SMILES OCC1=CC=CC=C1OC(F)(F)F

Specifications

Boiling Point 96°C to 98°C (12 mmHg)
Quantity 1 g
Formula Weight 192.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)benzyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.