missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 10748075
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10748075 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10748075 Supplier Thermo Scientific Maybridge Supplier No. SB00885DA
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 88-17-5
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007718
InChI Key VBLXCTYLWZJBKA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
PubChem CID 6922
IUPAC Name 2-(trifluoromethyl)aniline
SMILES C1=CC=C(C(=C1)C(F)(F)F)N

Specifications

CAS Min % 97.0
Density 1.2960g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula H2NC6H4CF3
Quantity 1 g
Formula Weight 161.13
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.