missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzonitrile, 97%

Product Code. 11404140
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404140 5 g 5g
11487137 10 g 10g
11497137 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404140 Supplier Thermo Scientific Alfa Aesar Supplier No. A14951.06
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 447-60-9 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 447-60-9
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001791
InChI Key SOZGHDCEWOLLHV-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
PubChem CID 67972
IUPAC Name 2-(trifluoromethyl)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C(F)(F)F

Specifications

Melting Point 17°C to 18°C
Density 1.294
Boiling Point 204°C to 206°C
Flash Point 90°C (194°F)
Refractive Index 1.4632
Quantity 5 g
UN Number UN3276
Beilstein 2048152
Formula Weight 171.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.