missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenol, 98%

Product Code. 11399599
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399599 1 g 1g
11402347 5 g 5g
11412347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399599 Supplier Thermo Scientific Alfa Aesar Supplier No. A14231.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 444-30-4
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002222
InChI Key ZOQOPXVJANRGJZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
PubChem CID 67958
IUPAC Name 2-(trifluoromethyl)phenol
SMILES C1=CC=C(C(=C1)C(F)(F)F)O

Specifications

Melting Point 45°C to 46°C
Boiling Point 147°C to 148°C
Flash Point 65°C (149°F)
Odor Phenol-like
Quantity 1 g
UN Number UN3261
Beilstein 1867917
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: .alpha.,.alpha.,.alpha.-Trifluoro-o-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.