missing translation for 'onlineSavingsMsg'
Learn More

2-(trifluoromethyl)phenyl isothiocyanate, Thermo Scientific™

Product Code. 10505265
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10505265 5 g 5g
11306346 10 g 10g
10397394 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10505265 Supplier Thermo Scientific Maybridge Supplier No. SB01593DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1743-86-8
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
InChI Key FCEKLQPJGXIQRY-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
PubChem CID 737162
IUPAC Name 1-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=S

Specifications

Density 1.346g/mL
Assay Percent Range 97%
Linear Formula CF3OC6H4NCS
Quantity 5 g
Formula Weight 203.19
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.