missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%

Product Code. 11460638
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460638 5 g 5g
11470638 25 g 25g
11480638 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460638 Supplier Thermo Scientific Alfa Aesar Supplier No. B25182.06
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3107-34-4 | C7H8ClF3N2 | 212.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3107-34-4
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD00102619
InChI Key ZUSWDTWYONAOPH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylhydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine hcl, 2-hydrazinobenzotrifluoride hcl, 2-hydrazinobenzotrifluoride hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine monohydrochloride, 1-2-trifluoromethyl phenyl hydrazine hydrochloride, n'-2-trifluoromethyl-phenyl-hydrazinium, chloride, 2-trifluoromethylphenylhydrazine hydrochloride
PubChem CID 12891828
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN.Cl

Specifications

Melting Point ∼235°C (decomposition)
Quantity 5 g
Beilstein 4582303
Sensitivity Hygroscopic
Formula Weight 212.6
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.