missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)thiophenol, 97%, Thermo Scientific™

Product Code. 11369945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11369945 1 g 1g
11379945 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11369945

Brand: Thermo Scientific Alfa Aesar L19913.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13333-97-6
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00068896
InChI Key XVGQHRKNXSUPEF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol
PubChem CID 2777889
IUPAC Name 2-(trifluoromethyl)benzenethiol
SMILES C1=CC=C(C(=C1)C(F)(F)F)S

Specifications

Density 1.35
Boiling Point 175°C to 177°C
Flash Point 68°C (154°F)
Refractive Index 1.499
Quantity 1 g
UN Number UN2810
Beilstein 2441965
Sensitivity Air Sensitive
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)thiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.