missing translation for 'onlineSavingsMsg'
Learn More

2-Trifluoromethylphenylboronic acid, 97%, ACROS Organics™

Product Code. 10501482 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10501482 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10501482 Supplier Thermo Scientific Acros Supplier No. 359745000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1423-27-4
CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H6BF3O2
MDL Number MFCD00236059
Quantity 500 mg
Synonym 2-trifluoromethyl phenylboronic acid, 2-trifluoromethylphenylboronic acid, 2-trifluoromethyl phenyl boronic acid, 2-trifluoromethyl benzeneboronic acid, 2-trifluoromethyl phenyl boranediol, o-trifluoromethylphenylboronic acid, 2-boronobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolueneboronic acid
InChI Key JNSBEPKGFVENFS-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C(F)(F)F)(O)O
IUPAC Name [2-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 189.928
PubChem CID 2734387
Formula Weight 189.93
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-Trifluoromethylphenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.