missing translation for 'onlineSavingsMsg'
Learn More

2-(Triphenylphosphoranylidene)propionaldehyde, 98%

Product Code. 10274864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274864 1 g Glass bottle 1g
10133054 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10274864 Supplier Thermo Scientific Acros Supplier No. 399110010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24720-64-7 | C21H19OP | 318.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24720-64-7
Molecular Formula C21H19OP
Molecular Weight (g/mol) 318.36
MDL Number MFCD00075596
InChI Key VHUQEFAWBCDBSC-UHFFFAOYSA-N
Synonym 2-triphenylphosphoranylidene propionaldehyde, 2-triphenylphosphoranylidene propanal, propanal, 2-triphenylphosphoranylidene, 1-formylethylidenetriphenylphosphorane, 2-triphenyl-??-phosphanylidene propanal, acmc-20aol9, 2-triphenylylidene propanal, 1-formylethylidene triphenyl phosphorane, alpha-formylethylidenetriphenylphosphorane, propanal,2-triphenylphosphoranylidene
PubChem CID 4280987
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)propanal
SMILES CC(C=O)=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 213.0°C to 217.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5)3P=C(CH3)COH
Quantity 1 g
Formula Weight 318.35
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 2-(Triphenylphosphoranylidene)propionaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Triphenylphosphoranylidene)propionaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.