missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylbenzoic acid, 96%

Product Code. 15464347
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464347 5 g 5g
15454347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464347 Supplier Thermo Scientific Alfa Aesar Supplier No. H31723.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27326-43-8 | C9H8O2 | 148.161 g/mol

As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As intermediate in orgaic syntheses. Phthalides were obtained in low yields by palladium-assisted cyclization of 2-vinylbenzoic acid and 2-(2-methyl-2-propenyl)- benzoic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27326-43-8
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD02066273
InChI Key XUDBVJCTLZTSDC-UHFFFAOYSA-N
PubChem CID 3015504
IUPAC Name 2-ethenylbenzoic acid
SMILES C=CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 94°C
Quantity 5 g
Synonym 2-vinylbenzoic acid, vinylbenzoic acid, benzoic acid, ethenyl, benzoic acid, 2-ethenyl, pubchem19668, benzoic acid,2-ethenyl, benzoic acid, 2-ethenyl-9ci
Formula Weight 148.16
Percent Purity 96%
Chemical Name or Material 2-Vinylbenzoic acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.