Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ Fixed Angle Rotors.
Brand:  Ohaus™ 30314898
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15846732
  • 102.00 € / Pack of 2
In Stock
to see stock.
Description
Description
  • Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
  • Suitable for use in the following Frontier™ rotors:
    • R-A4x85/12MI
    • R-A6x85/13MI
    • R-A6x85/14MI
Specifications
Specifications
RB
17 mm
Adapter, 2x15ml D17mm RB, 2/pk
2
2 x 15 mL
R-A4x85/12MI, R-A6x85/13MI, R-A6x85/14MI Rotors
2