Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ Fixed Angle Rotors.
102.00 €
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15896722
|
Ohaus™
30314893 |
1 x 15 mL (17 mm dia. RB) |
102.00 €
Pack of 2 |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15836732
|
Ohaus™
30314897 |
1 x 15 mL (17 mm dia. FA) |
102.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15846732
|
Ohaus™
30314898 |
2 x 15 mL |
102.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15806732
|
Ohaus™
30314894 |
1 x 30 mL |
102.00 €
Pack of 2 |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856732
|
Ohaus™
30314899 |
1 x 16 mL |
102.00 €
Pack of 2 |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15816732
|
Ohaus™
30314895 |
1 x 50 mL (29 mm dia. RB) |
102.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15826732
|
Ohaus™
30314896 |
1 x 50 mL (29 mm dia. FA) |
102.00 €
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
- Suitable for use in the following Frontier™ rotors:
- R-A4x85/12MI
- R-A6x85/13MI
- R-A6x85/14MI
Specifications
2 |