missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ Fixed Angle Rotors.
Brand: Ohaus™ 30314893
Additional Details : Weight : 1.00000kg
Description
- Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
- Suitable for use in the following Frontier™ rotors:
- R-A4x85/12MI
- R-A6x85/13MI
- R-A6x85/14MI
Specifications
RB | |
17 mm | |
Adapter, 1x15ml D17mm RB, 2/pk | |
2 |
1 x 15 mL (17 mm dia. RB) | |
Frontier Fixed-Angle Rotors | |
1 |