missing translation for 'onlineSavingsMsg'
Learn More

20% Biodiesel/80% High Cetane Diesel Fuel, v/v, VHG™

Product Code. 17922596
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922596 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17922596 Supplier VHG Labs Supplier No. VHGBDBLEND20P20
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

20% Biodiesel/80% High Cetane Diesel Fuel, v/v
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bio Diesel Fuel
CAS 870530-78-2
Analyte or Component Names Bio Diesel Fuel
Shipping Condition Room Temperature
Concentration 20%
Synonym Diesel fuel substitutes, biodiesel, B 10, B 10 biofuel, B 100, B 100 biofuel, B 20, B 20 biofuel, B 5, B 5 biofuel, B 50, B 50 biofuel, Biodiesel, Biofuels, biodiesel, Biodiesel fuel
IUPAC Name Biodiesel Fuel
Solvent or Matrix High Cetane Diesel Fuel/Biodiesel
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.