missing translation for 'onlineSavingsMsg'
Learn More

20 g/L Boric Acid Distillation Fluid, 50 x 10 mL, smart delivery PFA Tube, Dr. Ehrenstorfer

Product Code. 30327459
Change view
Click to view available options
Quantity:
50 x 10 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327459 50 x 10 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327459 Supplier Dr. Ehrenstorfer Supplier No. DREO000029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 x 10 mL
Concentration 20 g/L
Certifications/Compliance ISO 17025
Solvent or Matrix Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.