missing translation for 'onlineSavingsMsg'
Learn More

200.7, 6010 - Quality Control (QCS) Solution 2: Al, Fe, K, Li, Mo, Na, Sb, SiO2, Sn, Ti @ 100 μg/mL in 20% HCl, tr. HF, VHG™

Product Code. 17951606
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951606 500 mL 500mL
17981096 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17951606 Supplier VHG Labs Supplier No. VHG44QCS2Z500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

200.7, 6010 - Quality Control (QCS) Solution 2: Al, Fe, K, Li, Mo, Na, Sb, SiO2, Sn, Ti @ 100 μg/mL in 20% HCl, tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Antimony/Iron/Lithium/Molybdenum/Potassium/Silicon dioxide/Sodium/Tin/Titanium/Silicon Monoxide
CAS 7429-90-5, 7440-36-0, 7439-89-6, 7439-93-2, 7439-98-7, 9-7-7440
Methods ICP
Analyte or Component Names Aluminium/Antimony/Iron/Lithium/Molybdenum/Potassium/Silicon dioxide/Sodium/Tin/Titanium/Silicon Monoxide
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Aluminium 100 μg/mL/Antimony 100 μg/mL/Iron 100 μg/mL/Lithium 100 μg/mL/Molybdenum 100 μg/mL/Potassium 100 μg/mL/Silicon dioxide 100 μg/mL/Sodium 100 μg/mL/Tin 100 μg/mL/Titanium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.