missing translation for 'onlineSavingsMsg'
Learn More

200.7 Rev. 3.3, 6010A - Mixed Calibration Standard 1: Be @ 50; Mn @ 100; Cd, Zn @ 150; Se @ 200; Pb @ 500 μg/mL in 2% HNO3, VHG™

Product Code. 17920636
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17920636 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17920636 Supplier VHG Labs Supplier No. VHGMCS1100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

200.7 Rev. 3.3, 6010A - Mixed Calibration Standard 1: Be @ 50; Mn @ 100; Cd, Zn @ 150; Se @ 200; Pb @ 500 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beryllium/Manganese/Cadmium/Zinc/Selenium/Lead
CAS 7440-41-7, 7439-96-5, 7440-43-9, 7440-66-6, 7782-49-2, 7439-92-1
Methods ICP/ICP-MS
Analyte or Component Names Beryllium/Manganese/Cadmium/Zinc/Selenium/Lead
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Beryllium 50.01 μg/mL/Manganese 100 μg/mL/Cadmium 149.9 μg/mL/Zinc 150 μg/mL/Selenium 200.1 μg/mL/Lead 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.