missing translation for 'onlineSavingsMsg'
Learn More
Learn More
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 μg/mL in 20% HCl, tr. HF, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 μg/mL in 20% HCl, tr. HF
Specifications
Specifications
| Chemical Name or Material | Tin/Antimony/Lithium/Aluminium/Iron/Molybdenum/Silicon dioxide/Sodium/Titanium/Silicon Monoxide |
| CAS | 7440-31-5, 7440-36-0, 7439-93-2, 7429-90-5, 7439-89-6, 7439-98-7 |
| Methods | ICP |
| Analyte or Component Names | Tin/Antimony/Lithium/Aluminium/Iron/Molybdenum/Silicon dioxide/Sodium/Titanium/Silicon Monoxide |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HCl |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Tin 200 μg/mL/Antimony 500 μg/mL/Lithium 500 μg/mL/Aluminium 1000 μg/mL/Iron 1000 μg/mL/Molybdenum 1000 μg/mL/Silicon dioxide 1000 μg/mL/Sodium 1000 μg/mL/Titanium 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction