missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 203.2 dia. x 50.8mmH Stainless Steel Test Sieve, ISO Certificate, Pore Size: 10mm and Larger

Brand:  RETSCH 60.149.019000

Additional Details : Weight : 0.35000kg

 View more versions of this product

Product Code. 10024971

  • 168.00 € / Each

Estimated Shipment: 23-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
50.8mm
For separation, fractioning, particle size determination
203.2mm
19mm
ISO 3310/1
Shaker AS 300
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers