missing translation for 'onlineSavingsMsg'
Learn More

ZT 52656A hydrochloride, MedChemExpress

Product Code. 16430426
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430426 20 mg 20mg
16460789 1 mg 1mg
16453438 10 mg 10mg
16406167 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16430426 Supplier Medchem Express Supplier No. HY101582
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye.

ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye.

Specifikationer

Chemical Name or Material ZT 52656A hydrochloride
Physical Form Solid
CAS 115730-24-0
Color Off-White
Quantity 20 mg
Purity Grade Notes Research
Molecular Formula C19H26ClF3N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 35.71 mg/mL (91.36 mM; Need ultrasonic)
SMILES O=C(CC1=CC=C(C=C1)C(F)(F)F)N2CCCCC2CN3CCCC3.Cl
Molecular Weight (g/mol) 390.87
Formula Weight 390.87
Percent Purity 99.76%
Grade Research
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.