missing translation for 'onlineSavingsMsg'
Learn More

Samuraciclib hydrochloride, MedChemExpress

Product Code. 16412906
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16412906 25 mg 25mg
16474936 100 mg 100mg
16446627 5 mg 5mg
16440267 50 mg 50mg
16413908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16412906 Supplier Medchem Express Supplier No. HY103712A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Samuraciclib hydrochloride (CT7001 hydrochloride) is a potent, selective, ATP-competitive and orally active CDK7 inhibitor, with an IC50 of 41 nM.

Samuraciclib hydrochloride displays 45-, 15-, 230- and 30-fold selectivity over CDK1, CDK2 (IC50 of 578 nM), CDK5 and CDK9, respectively. Samuraciclib hydrochloride inhibits the growth of breast cancer cell lines with GI50 values between 0.2-0.3 μM. Samuraciclib hydrochloride has anti-tumor effects.

Specifications

Chemical Name or Material Samuraciclib hydrochloride
Physical Form Powder
CAS 1805789-54-1
Color White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C22H31ClN6O
For Use With (Application) Cancer-Kinase/protease
Synonym CT7001 hydrochlorideICEC0942 hydrochloride
Solubility Information H2O : 55 mg/mL (127.62 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (116.02 mM; Need ultrasonic)
SMILES O[C@H]1CNCC[C@@H]1CNC2=NC3=C(C(C)C)C=NN3C(NCC4=CC=CC=C4)=C2.[H]Cl
Molecular Weight (g/mol) 430.97
Formula Weight 430.97
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.