missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase-13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76812-0.025mg
This item is not returnable.
View return policy
Description
Caspase-13 Polyclonal specifically detects Caspase-13 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Caspase-4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| apoptotic cysteine protease Mih1/TX, CASP-4, caspase 4, apoptosis-related cysteine peptidase, caspase 4, apoptosis-related cysteine protease, caspase-4, EC 3.4.22, EC 3.4.22.57, ICE(rel)II, ICE(rel)-II, ICEREL-II, ICH2, ICH-2, Mih1/TX, Protease ICH-2, Protease TX, TX | |
| Rabbit | |
| 43 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer, Caspases | |
| 338039 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| O75601 | |
| CASP4 | |
| Antibody was raised against a 15 amino acid peptide near the amino terminus of bovine Caspase-13. The immunogen is located within amino acids 80 - 130 of Caspase-13. Amino Acid Squence: ETVAGPDESVGSAAT | |
| Ion exchange chromatography | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction