missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Otx2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 47 publications
Brand: R&D Systems AF1979-SP
This item is not returnable.
View return policy
Description
Otx2 Polyclonal specifically detects Otx2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| Otx2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5015 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P32243 | |
| OTX2 | |
| E. coli-derived recombinant human Otx2 Met1-Leu289 Accession # P32243 | |
| 25 μg | |
| Primary | |
| Detects human Otx2 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction