missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat SMOC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5550-SP
This item is not returnable.
View return policy
Description
SMOC-1 Polyclonal specifically detects SMOC-1 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SMOC-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Secreted modular calcium-binding protein 1, SMOC1, SMOC-1, SPARC related modular calcium binding 1, SPARC-related modular calcium-binding protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 64093 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAD41590 | |
| SMOC1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SMOC-1 His26-Val452 Accession # AAD41590 | |
| 25 μg | |
| Primary | |
| Detects mouse and rat SMOC-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) SMOC-1 is observed and less than 1% cross-reactivity with recombinant mouse SMOC-2 and rhSMOC-2 is observed. | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction