missing translation for 'onlineSavingsMsg'
Learn More

Mouse ULBP-1/MULT-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18737069
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18737069 25 μg 25µg
18129390 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18737069 Supplier R&D Systems Supplier No. MAB2588SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 12 publications

ULBP-1 Monoclonal specifically detects ULBP-1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULBP-1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 237104
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1
Gene Symbols ULBP1
Host Species Rat
Immunogen BaF3 mouse pro-B cell line transfected with mouse ULBP-1MULT-1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80329
Test Specificity Detects mouse ULBP-1MULT-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.