missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
HOXB9 Antibody [Unconjugated], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43680-25ul
This item is not returnable.
View return policy
Description
HOXB9 Polyclonal antibody specifically detects HOXB9 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| HOXB9 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| homeo box 2E, homeo box B9, homeobox B9, Homeobox protein Hox-2.5, Homeobox protein Hox-2E, homeobox protein Hox-B9, HOX-2.5, HOX2EHOX2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SISGTLSSYYVDSIISHESEDAPPAKFPSGQYASSRQPGHAEHLEFPSCSFQPKAPVFGASWAPLSPHASGSLPSVYHPYIQPQGVPPAESRYLRTWLEP | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3219 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction