missing translation for 'onlineSavingsMsg'
Learn More
Learn More
28 Elements Solution, 0.1% C6H4O6 matrix, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Specifications
Specifications
| Solution Contains | Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, K, Li, Mg, Mn, Mo, Na, Ni, Pb Sb Se Sr, Ti, Tl, V, Zn for ICP 10mg/L of each 5% HNO3, 0.1% C6H4O6 matrix |
| Concentration | 10 mg/L |
| Quantity | 100 mL |
Safety and Handling
Product Identifier
- Multi-element Standard Solution for ICP. 28 Components in HNO3 5% ,C4H6O6 0.1% (ME/1001/)
- Danger
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Partager une correction de contenu