missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (025), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89232CL1
This item is not returnable.
View return policy
Description
2B4/CD244/SLAMF4 Monoclonal antibody specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot,Sandwich ELISA Capture
Specifications
| 2B4/CD244/SLAMF4 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human 2B4/CD244/SLAMF4 (Accession#: NP_057466.1; Met1-Arg221). | |
| 0.1 mL | |
| Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells | |
| 51744 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot, Sandwich ELISA Capture | |
| 025 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction