missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 26/77 film, 3 ports, Luer Lock insert and body connectors with reducers, injection site

Product Code. 17879776
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1 L
2 L
Unit Size:
Pack of 30
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17879776 2 L Pack of 30
17805736 100 mL Pack of 50
17825736 250 mL Pack of 30
17895726 500 mL Pack of 50
17855736 1 L Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17879776 Supplier Thermo Scientific™ Supplier No. SS00033I
  • ONLINE EXCLUSIVE PRICE
    1450.00 €
    1235.36 € / Pack of 30
    Save 214.64 €
    14% Off
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

The Thermo Scientific™ BioProcess Container (BPC) 2D System is composed of Thermo Scientific™ ASI™ 26/77 Polyethylene Film, a single-web, multi-layer 12.5 mL polyethylene film.

Features

  • Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
  • Constructed in an ISO 7 clean room under cGMP conditions

Applications

  • Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
  • Preparing and storing buffer, culture media, and process liquids
  • Collecting chromatography feeds and fractions
  • Harvesting, storing, and transporting bulk drug product and drug precursors

Customize standard BPC systems for optimized single-use technology
Choose from the industry's largest component library, with over 2,000 unique components that include:

  • Fittings – straight, reducing, elbow, T-style, X-style, cross-style
  • Connectors –quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
  • Filters (used for bioburden reduction ) –sterilization grade filters, vent filters
  • Tubing –thermoplastic elastomers, platinum-cured silicone
  • Sample ports –septum-style ports, needle-free ports
  • Clamps –sliders, pinch clamps, crimp bands

BPC port specifications:

  • Line 1: Luer lock insert connection with 4.8 mm (3/16 in.) barb, cap, slide clamp, and 4.8 x 4.1 mm (3/16 x 5/32 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
  • Line 2: Luer lock body connection with 0.56 cm (0.22 in.) barb, plug, slide clamp, and 0.48 x 0.41 cm (0.12 x 0.16 in.) reducer. Tubing: TPE 374, length: 30 cm (12 in.), ID x OD: 3.2 x 6.4 mm (0.13 x 0.25 in.)
  • Line 3: Tubing: EVA, injection port
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2 L
Material Polyethylene
Product Type BioProcess Container
Content And Storage Store at room temperature.
Dimensions 30.5 x 20.3 cm (12.0 x 8.0 in.)
Outer Packaging One 4-mil. poly bag
Quantity 50/Cs.
Sterilization Method Irradiated
Shipping Condition Room Temperature
Product Line Labtainer
Port Detail 3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site)
Film Description ASI 26/77
Tubing Style TPE and EVA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.