missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic Acid, TRC

Product Code. 30240975
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240975 1 g 1g
30240609 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240975 Supplier Toronto Research Chemicals Supplier No. TRCB2035201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38489-76-8
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
Synonym (E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic Acid, (2E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic Acid, (E)-3,4-(Methylenedioxy)cinnamic Acid, (E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic Acid, (E)-3-(3,4-Methylenedioxyphenyl)acrylic Acid, 3-(3,4-Methylenedioxyphenyl)-2E-propenoic Acid, trans-3,4-(Methylenedioxy)cinnamic Acid, trans-3-(3,4-Methylenedioxyphenyl)-2-propenoic Acid
IUPAC Name (E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
SMILES OC(=O)\C=C\c1ccc2OCOc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H8O4/c11-10(12)4-2-7-1-3-8-9(5-7)14-6-13-8/h1-5H,6H2,(H,11,12)/b4-2+
Formula Weight 192.0423
Chemical Name or Material (2E)-3-(1,3-Benzodioxol-5-yl)-2-propenoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.