missing translation for 'onlineSavingsMsg'
Learn More

(2E)-3-Phenyl-2-propenoic Acid, TRC

Product Code. 30011521
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011521 10 g 10g
30010588 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011521 Supplier Toronto Research Chemicals Supplier No. TRCP33618510G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-10-3
Molecular Formula C9 H8 O2
Molecular Weight (g/mol) 148.16
Synonym 2-Propenoic acid, 3-phenyl-, (2E)- (9CI, ACI), (2E)-3-Phenyl-2-propenoic acid (ACI), 2-Propenoic acid, 3-phenyl-, (E)- (ZCI), Cinnamic acid, (E)- (8CI), (2E)-2-Phenyl-2-propenoic acid, (2E)-3-Phenylprop-2-enoic acid, (2E)-Cinnamic acid, (E)-3-Phenyl-2-propenoic acid, (E)-3-Phenylacrylic acid, (E)-3-Phenylacrylic acid, (E)-3-Phenylprop-2-enoic acid, (E)-Cinnamic acid, 3-Phenyl-(E)-2-propenoic acid, NSC 44010, trans-3-Phenyl-2-propenoic acid, trans-3-Phenylacrylic acid, trans-Cinnamic acid, trans-Cinnamic acid, trans-β-Carboxystyrene
IUPAC Name (E)-3-phenylprop-2-enoic acid
SMILES OC(=O)\C=C\c1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H8O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H,(H,10,11)/b7-6+
Formula Weight 148.0524
Chemical Name or Material trans-Cinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.