missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid, 97%

Product Code. 15400136
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400136 250 mg 250mg
15490126 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15400136 Supplier Thermo Scientific Alfa Aesar Supplier No. H52806.MD
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 73397-28-1 | C12H23NO5 | 261.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 73397-28-1
Molecular Formula C12H23NO5
Molecular Weight (g/mol) 261.318
MDL Number MFCD04972275
InChI Key DJZCWTDKDFJARG-DTWKUNHWSA-N
Synonym boc-2r,3s-3-amino-2-hydroxy-5-methylhexanoicacid, 2r,3s-3-boc-amino-2-hydroxy-5-methylhexanoic acid, n-boc-2r,3s-2-hydroxy-3-amino-5-methylhexanoic acid, 2r,3s-3-tert-butoxycarbonyl amino-2-hydroxy-5-methylhexanoic acid, 2r,3s-2-hydroxy-3-tert-butoxycarbonylamino-5-methylhexanoic acid
PubChem CID 10377964
IUPAC Name (2R,3S)-2-hydroxy-5-methyl-3-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)CC(C(C(=O)O)O)NC(=O)OC(C)(C)C

Especificaciones

Quantity 250 mg
Formula Weight 261.32
Percent Purity 97%
Chemical Name or Material (2R,3S)-3-(Boc-amino)-2-hydroxy-5-methylhexanoic acid

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.