missing translation for 'onlineSavingsMsg'
Learn More

(2R,3S)-N-Benzoyl-3-phenyl Isoserine Ethyl Ester, TRC

Product Code. 30137383
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137383 500 mg 500mg
17925065 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137383 Supplier Toronto Research Chemicals Supplier No. TRCB208555500MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153433-80-8
Molecular Formula C18 H19 N O4
Molecular Weight (g/mol) 313.35
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, ethyl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, ethyl ester, [R-(R*,S*)]-, Ethyl (2R,3S)-3-(benzoylamino)-2-hydroxybenzenepropanoate, (2R,3S)-N-Benzoyl-3-phenylisoserine ethyl ester, Ethyl (2R,3S)-3-Benzamido-2-hydroxy-3-phenylpropanoate
IUPAC Name ethyl (2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoate
SMILES CCOC(=O)[C@H](O)[C@@H](NC(=O)c1ccccc1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H19NO4/c1-2-23-18(22)16(20)15(13-9-5-3-6-10-13)19-17(21)14-11-7-4-8-12-14/h3-12,15-16,20H,2H2,1H3,(H,19,21)/t15-,16+/m0/s1
Formula Weight 313.1314
Percent Purity >95%
Chemical Name or Material N-Benzyloxymethylphenylisoserine ethylester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.