missing translation for 'onlineSavingsMsg'
Learn More

2R,4R-Sacubitril, MedChemExpress

Product Code. 16461989
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16461989

Brand: Medchem Express HY78846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2R,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.

2R,4R-Sacubitril is the impurity of Sacubitril. Sacubitril is approved by the Food and Drug Administration for use in combination with valsartan for the treatment of patients with heart failure.

Specifications

Chemical Name or Material 2R,4R-Sacubitril
Physical Form Solid
CAS 766480-48-2
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C24H29NO5
For Use With (Application) COVID-19-immunoregulation
SMILES [H]N(C(CCC(O)=O)=O)[C@@H](CC(C=C1)=CC=C1C2=CC=CC=C2)C[C@H](C(OCC)=O)C
Molecular Weight (g/mol) 411.49
Formula Weight 411.49
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.