missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Benzothiazol-2-yl)-4-methylthiophen-2-amine, Thermo Scientific™

Product Code. 10563202
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10563202 1 g 1g
10427184 5 g 5g
10600106 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10563202 Supplier Thermo Scientific Maybridge Supplier No. GK03755DA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 232941-00-3
Molecular Formula C12H10N2S2
Molecular Weight (g/mol) 246.35
MDL Number MFCD02179980
InChI Key YXXFAQKMMOVVIA-UHFFFAOYSA-N
Synonym 3-1,3-benzothiazol-2-yl-4-methylthiophen-2-amine, 2-2-amino-4-methylthien-3-yl benzothiazole, 3-benzo d thiazol-2-yl-4-methylthiophen-2-amine, 2-thiophenamine,3-2-benzothiazolyl-4-methyl, maybridge1_003812, 3-benzothiazol-2-yl-4-methyl-2-thienylamine, 2-2-amino-4-methylthien-3-yl-1,3-benzothiazol, 2-2-amino-4-methylthien-3-yl-1,3-benzothiazole, 3-1,3-benzothiazol-2-yl-4-methyl-2-thiophenamine, 3-1,3-benzothiazol-2-yl-4-methyl-thiophen-2-amine
PubChem CID 2735376
IUPAC Name 3-(1,3-benzothiazol-2-yl)-4-methylthiophen-2-amine
SMILES CC1=CSC(N)=C1C1=NC2=CC=CC=C2S1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 246.36
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Benzothiazol-2-yl)-4-methylthiophen-2-amine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.