missing translation for 'onlineSavingsMsg'
Learn More

3-(1,3-Thiazol-2-yl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10708465
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10708465 1 g Amber Glass Bottle 1g
10265764 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10708465 Supplier Thermo Scientific Maybridge Supplier No. CC36501DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 847956-27-8
Molecular Formula C10H7NO2S
Molecular Weight (g/mol) 205.231
MDL Number MFCD06797800
InChI Key BLFSSABBFANDKK-UHFFFAOYSA-N
PubChem CID 21075087
IUPAC Name 3-(1,3-thiazol-2-yl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=NC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-1,3-thiazol-2-yl benzoic acid, 3-thiazol-2-yl-benzoic acid, 3-thiazol-2-yl benzoic acid, benzoic acid,3-2-thiazolyl, benzoic acid, 3-2-thiazolyl, 3-1,3-thiazol-2-yl benzoicacid, 2-3-carboxyphenyl-1,3-thiazole
Formula Weight 205.24
Percent Purity 97%
Chemical Name or Material 3-(1, 3-Thiazol-2-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.